Tubing
: Agilent
Copper and stainless steel tubing for gas chromatography (GC) applications.
- Copper can be used for most applications, except those requiring the highest purity gas delivery system
- Stainless steel and UltiMetal Plus tubing are appropriate for more demanding applications
These high-quality copper, stainless steel, and UltiMetal Plus tubing are manufactured and cleaned for chromatographic use.
Frequently Bought Together