Order Entry
Ireland
ContactUsLinkComponent
FcR-IIIA-5PW, TSKgel®
FcR-IIIA-5PW, TSKgel®
Catalog # TOYO0023532
Supplier:  TOSOH Bioscience
FcR-IIIA-5PW, TSKgel®
Catalog # TOYO0023532
Supplier:  TOSOH Bioscience

Specifications

  • Column Format:
    Semi-preparative
  • Ø int.:
    7,80 mm
  • Particle Size:
    10 µm
  • Max. pressure:
    1 MPa
  • Description:
    TSKgel FcR-IIIA-5PW
  • Length:
    75 mm
  • pH range:
    4,0 - 8,0
  • Pk:
    1

Specifications

About this item

TSKgel FcR-IIIA-5PW is a semi-preparative affinity column which immobilises the recombinant FcγRIIIA ligand bonded to pourous 10 µm polymethacrylate particles which can load up to 5 mg of mAb.

  • Semi-preparative affinity column for collecting fractions with different FcR affinities to characterise them
  • Temperature range 15 to 25 °C

Isolate antibodies according to FcR Affinity from a heterogeneous sample. Expression of antibodies typically leads to an antibody mixture with different affinities to the FcγIIIa receptor. The TSKgel FcR-IIIA-5PW column can be loaded with 5 mg of antibody sample and resolves three affinity fractions. These can be employed to analyse structural and functional differences linked to differences in FcR affinity.

Delivery: Shipping solvent: 0,65 mmol citric acid, 9,35 mmol/L trisodium citrate containing 0,025 ProClin® 300, pH 6,5.
Packaging: Each package includes the related HPLC column as well as the operating conditions and specification sheet.