Order Entry
Ireland
ContactUsLinkComponent
alpha-Biotin-omega-carboxysuccinimidyl ester poly(ethyleneglycol), M.W. PEG molecular weight 3,000 daltons
Irritant
alpha-Biotin-omega-carboxysuccinimidyl ester poly(ethyleneglycol), M.W. PEG molecular weight 3,000 daltons
Catalog # APOSBIP1056-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
alpha-Biotin-omega-carboxysuccinimidyl ester poly(ethyleneglycol), M.W. PEG molecular weight 3,000 daltons
Catalog # APOSBIP1056-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient