Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-IL-12p40 Rat Antibody
Anti-IL-12p40 Rat Antibody
Anti-IL-12p40 Rat Antibody
Catalog # APOSBITP2016-1MG
Supplier:
Apollo Scientific
Expand image
Anti-IL-12p40 Rat Antibody
Catalog # APOSBITP2016-1MG
Supplier:
Apollo Scientific
Specifications
Antigen name:
Interleukin-12 p40
Antigen symbol:
IL-12p40
Host:
Rat
Reactivity:
Mouse
Pk:
1 mg
Specifications
About this item
Product Details
Product Details