Order Entry
Ireland
ContactUsLinkComponent
Recombinant Treponema pallidum TmpA (partial)
Recombinant Treponema pallidum TmpA (partial)
Catalog # APOSBITP1862-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Treponema pallidum TmpA (partial)
Catalog # APOSBITP1862-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    Treponema TmpA
  • Protein/peptide name:
    Treponema pallidum TmpA (partial)

Specifications

About this item