0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Proteins and Peptides
Recombinant Cytomegalo Virus pP52 (UL44)
Recombinant Cytomegalo Virus pP52 (UL44)
APOSBITP1842-1MG
:
Apollo Scientific
:
Recombinant Cytomegalo Virus pP52 (UL44)
APOSBITP1842-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
CMV Pp52
Protein/peptide name:
Cytomegalo Virus pP52 (UL44)