Order Entry
Ireland
ContactUsLinkComponent
Recombinant Hepatitis B Virus Core (1-183)
Recombinant Hepatitis B Virus Core (1-183)
Catalog # APOSBITP1821-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis B Virus Core (1-183)
Catalog # APOSBITP1821-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HBcAg
  • Protein/peptide name:
    Hepatitis B Virus Core (1-183)

Specifications

About this item