Order Entry
Ireland
ContactUsLinkComponent
Soybean recombinant P34 Protein
Soybean recombinant P34 Protein
Catalog # APOSBITP1722-1MG
Supplier:  Apollo Scientific
CAS Number:  
Soybean recombinant P34 Protein
Catalog # APOSBITP1722-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Soy bean
  • Protein synonyms:
    Soy Bean P34
  • Protein/peptide name:
    P34 Protein

Specifications

About this item