0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Proteins and Peptides
Human recombinant GH
Human recombinant GH
Catalog # APOSBITP1067-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Human recombinant GH
Catalog # APOSBITP1067-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Protein synonyms:
Growth Hormone
Protein/peptide name:
GH
Specifications
About this item
Product Details
Product Details