6-deoxy-6-bromo-heptakis(6-deoxy-6-(2-carboxyethyl)thio)-gamma-cyclodextrin sodium salt
Catalog # APOSBICY1067-5MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
Storage Temperature:Ambient