Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-deoxy-6-[(5/6)-fluoresceinylthioureido]-RAMEB
6-deoxy-6-[(5/6)-fluoresceinylthioureido]-RAMEB
6-deoxy-6-[(5/6)-fluoresceinylthioureido]-RAMEB
6-deoxy-6-[(5/6)-fluoresceinylthioureido]-RAMEB
Catalog # APOSBICY1045-5MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
6-deoxy-6-[(5/6)-fluoresceinylthioureido]-RAMEB
Catalog # APOSBICY1045-5MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pack type:
Bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
Storage Temperature:
Ambient
Product Details