Order Entry
Ireland
ContactUsLinkComponent
1,2-Dibromo-4-fluorobenzene 98%
1,2-Dibromo-4-fluorobenzene 98%
Catalog # APOSPC9619-100G
Supplier:  Apollo Scientific
CAS Number:  
1,2-Dibromo-4-fluorobenzene 98%
Catalog # APOSPC9619-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :253.9 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:2369-37-1
Formula:C₆H₃Br₂F
MDL Number:MFCD00070780