7-{1-[3-(Trifluoromethyl)phenoxy]ethyl}[1,2,4]triazolo[1,5-a]pyrimidin-2-amine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306979-68-0
:
Formula:C₁₄H₁₂F₃N₅O
MDL Number:MFCD00243589
:323.28 g/mol
Melting Pt:182…184 °C
Storage Temperature:Ambient
Frequently Bought Together