Order Entry
Ireland
ContactUsLinkComponent
5-Fluoro-2-methyl-1-indanone 95%
5-Fluoro-2-methyl-1-indanone 95%
Catalog # APOSPC905377-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Fluoro-2-methyl-1-indanone 95%
Catalog # APOSPC905377-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.18 g/mol
Storage Temperature:Ambient
CAS Number:41201-58-5
Formula:C₁₀H₉FO
MDL Number:MFCD07787592