
3-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-5-ylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:209224-91-9
:
Formula:C₁₀H₈F₃N₃
MDL Number:MFCD04110829
:227.19 g/mol
Melting Pt:130…132 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together