Order Entry
Ireland
ContactUsLinkComponent
2,3,4,5,6-Pentafluorobenzyl chloroformate 97%
Corrosive
Irritant
2,3,4,5,6-Pentafluorobenzyl chloroformate 97%
Catalog # APOSPC8579-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2,3,4,5,6-Pentafluorobenzyl chloroformate 97%
Catalog # APOSPC8579-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :260.55 g/mol
Boiling Pt:116 °C
Storage Temperature:Ambient
UN:3277
ADR:6.1+8,II
 :GHS05
 :GHS07
CAS Number:53526-74-2
Formula:C₈H₂ClF₅O₂
MDL Number:MFCD00798136