Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5,6-Trifluoropyrimidine
4,5,6-Trifluoropyrimidine
4,5,6-Trifluoropyrimidine
4,5,6-Trifluoropyrimidine
APOSPC7842-250MG
:
Apollo Scientific
:
4,5,6-Trifluoropyrimidine
APOSPC7842-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
134.06 g/mol
:
83…85 °C
:
1,4609 g/cm³ (25 °C)
:
98 °C
:
GHS07
:
Ambient
:
17573-78-3
:
C₄HF₃N₂
:
MFCD05662363