Order Entry
Ireland
ContactUsLinkComponent
2,4,6-Trifluoropyrimidine 97%
2,4,6-Trifluoropyrimidine 97%
Catalog # APOSPC7834-5G
Supplier:  Apollo Scientific
CAS Number:  
2,4,6-Trifluoropyrimidine 97%
Catalog # APOSPC7834-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :134.06 g/mol
Boiling Pt:100…102 °C
Flash Pt:>100 °C
Storage Temperature:Ambient
CAS Number:696-82-2
Formula:C₄HF₃N₂
MDL Number:MFCD00039705