(2,4,6-Trifluorophenyl)acetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:220227-80-5
:
Formula:C₈H₄F₃N
MDL Number:MFCD00061221
:171.12 g/mol
Boiling Pt:103 °C/3mmHg
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
Frequently Bought Together