2-Bromo-4-chloro-1-fluorobenzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1996-30-1
:
Formula:C₆H₃BrClF
MDL Number:MFCD00672934
:209.44 g/mol
Boiling Pt:180 °C
Density:1,719 g/cm³ (25 °C)
Flash Pt:85 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together