2-Bromo-4-fluorobenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:36282-26-5
:
Formula:C₇H₃BrFN
MDL Number:MFCD00672924
:200.01 g/mol
Melting Pt:77…78 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together