2,4,6-Trifluorobenzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:28314-80-9
:
Formula:C₇H₃F₃O₂
MDL Number:MFCD00042398
:176.09 g/mol
Melting Pt:143…145 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together