2,4,6-Trifluorobenzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:58551-83-0
:
Formula:C₇H₃F₃O
MDL Number:MFCD00061197
:160.1 g/mol
Boiling Pt:165 °C
Melting Pt:63…68 °C
Density:1,407 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together