2,4,6-Trifluoroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:363-81-5
:
Formula:C₆H₄F₃N
MDL Number:MFCD00007650
:147.1 g/mol
Boiling Pt:57 °C/22mmHg
Melting Pt:33…37 °C
Flash Pt:58 °C
EINECS:206-660-8
UN:1325
ADR:4.1,III
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together