3-Bromo-2-fluorobenzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:840481-82-5
:
Formula:C₇H₃BrFN
MDL Number:MFCD06657980
:200.01 g/mol
Melting Pt:37…39 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together