3-Oxo-N-(3-(trifluoromethyl)phenyl)butanamide
Catalog # APOSPC6299-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:245.2 g/mol
Storage Temperature:Ambient
CAS Number:785-74-0
Formula:C₁₁H₁₀F₃NO₂
MDL Number:MFCD00201876