
4-(4-Trifluoromethylphenyl)thiazol-2-ylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:105512-79-6
:
Formula:C₁₀H₇F₃N₂S
MDL Number:MFCD03094298
:244.24 g/mol
Melting Pt:206…208 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together