
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol 95+%
Catalog # APOSPC53582-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.10 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:368-54-7
Formula:C₅H₃F₃N₂OS
MDL Number:MFCD00112978