Order Entry
Ireland
ContactUsLinkComponent
Methyl-2-amino-3-fluorobenzoate
Irritant
Methyl-2-amino-3-fluorobenzoate
Catalog # APOSPC53196-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-2-amino-3-fluorobenzoate
Catalog # APOSPC53196-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :169.1530 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:144851-82-1
Formula:C₈H₈FNO₂
MDL Number:MFCD11870179