1-[2-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethyl-5-cyano-1,2,3,4-tetrahydropyrimidine-2,4-dione
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338409-11-3
:
Formula:C₁₃H₉ClF₃N₅O₂
MDL Number:MFCD00172995
:359.70 g/mol
Melting Pt:215…217 °C
Storage Temperature:Ambient
Frequently Bought Together