4-Fluoro-o-phenylenediamine, Technical Grade
Catalog # APOSPC48407-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:126.13 g/mol
Melting Pt:94…98 °C
Flash Pt:>110 °C
EINECS:206-691-7
:GHS07
Storage Temperature:Refrigerator
CAS Number:367-31-7
Formula:C₆H₇FN₂
MDL Number:MFCD00042228