
4-(Difluoromethoxy)benzylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:177842-14-7
:
Formula:C₈H₉F₂NO
MDL Number:MFCD00236231
:173.16 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together