Order Entry
Ireland
ContactUsLinkComponent
tert-Butyl-4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate
tert-Butyl-4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate
Catalog # APOSPC430278-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate
Catalog # APOSPC430278-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :341.81 g/mol
Storage Temperature:Ambient
CAS Number:1228631-50-2
Formula:C₁₇H₂₁ClFNO₃
MDL Number:MFCD16618544