
4-Bromo-2-fluoro-5-methylbenzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:415965-24-1
:
Formula:C₈H₆BrFO₂
MDL Number:MFCD08458245
:233.0300 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together