4-Fluoro-2-trifluoromethoxy-DL-phenylalanine
Catalog # APOSPC302877-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:267.18 g/mol
Storage Temperature:Ambient
CAS Number:1435806-18-0
Formula:C₁₀H₉F₄NO₃
MDL Number:MFCD19687192