5-Fluoro-2-methoxy-DL-phenylalanine
Catalog # APOSPC302622-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.21 g/mol
Melting Pt:251 °C
Storage Temperature:Ambient
CAS Number:603105-80-2
Formula:C₁₀H₁₂FNO₃
MDL Number:MFCD05227952