5-Oxo-1-[4-(trifluoromethoxy)phenyl]pyrrolidine-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:874765-55-6
:
Formula:C₁₂H₁₀F₃NO₄
MDL Number:MFCD01569312
:289.21 g/mol
Storage Temperature:Ambient
Frequently Bought Together