4-Fluoroveratrole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:398-62-9
:
Formula:C₈H₉FO₂
MDL Number:MFCD00012201
:156.16 g/mol
Boiling Pt:120…123 °C/45mmHg
Density:1,171 g/cm³ (25 °C)
Flash Pt:91 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together