Order Entry
Ireland
ContactUsLinkComponent
5-Oxo-1-[4-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid 95%
Irritant
5-Oxo-1-[4-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid 95%
Catalog # APOSPC2954-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Oxo-1-[4-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid 95%
Catalog # APOSPC2954-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :273.21 g/mol
Melting Pt:110…112 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:2357-27-9
Formula:C₁₂H₁₀F₃NO₃
MDL Number:MFCD00665831