Order Entry
Ireland
ContactUsLinkComponent
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-piperidine-4-ol
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-piperidine-4-ol
Catalog # APOSPC2952-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-piperidine-4-ol
Catalog # APOSPC2952-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :280.68 g/mol
Melting Pt:74…75 °C
Storage Temperature:Ambient
CAS Number:306976-40-9
Formula:C₁₁H₁₂ClF₃N₂O
MDL Number:MFCD00202495