Order Entry
Ireland
ContactUsLinkComponent
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxamide
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxamide
Catalog # APOSPC2949-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxamide
Catalog # APOSPC2949-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :307.71 g/mol
Melting Pt:187…188 °C
Storage Temperature:Ambient
CAS Number:338780-66-8
Formula:C₁₂H₁₃ClF₃N₃O
MDL Number:MFCD00172305