2,4,6-Trifluorobenzylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:214759-21-4
:
Formula:C₇H₆F₃N
MDL Number:MFCD00236317
:161.13 g/mol
Boiling Pt:165 °C
Density:1,32 g/cm³ (25 °C)
Flash Pt:63 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together