1,2-Dichloro-3-fluorobenzene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:36556-50-0
:
Formula:C₆H₃Cl₂F
MDL Number:MFCD00060647
:164.9924 g/mol
Boiling Pt:80 °C/22mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together