1-Bromo-3-chloro-2-fluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:144584-65-6
:
Formula:C₆H₃BrClF
MDL Number:MFCD03412182
:209.44 g/mol
Boiling Pt:88…92 °C/24mmHg
Melting Pt:43 °C
Flash Pt:74 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together