5-Bromo-2-fluorobenzonitrile 98%
Catalog # APOSPC1420M-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:200.01 g/mol
Melting Pt:74…77 °C
:GHS07
Storage Temperature:Ambient
CAS Number:179897-89-3
Formula:C₇H₃BrFN
MDL Number:MFCD00143424