4-Bromo-2-fluoroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:367-24-8
:
Formula:C₆H₅BrFN
MDL Number:MFCD00010221
:190.01 g/mol
Boiling Pt:103…108 °C/13mmHg
Melting Pt:38…42 °C
Flash Pt:104 °C
Storage Temperature:Ambient
EINECS:206-685-4
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together