1-Bromo-3-chloro-5-fluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:33863-76-2
:
Formula:C₆H₃BrClF
MDL Number:MFCD00070747
:209.44 g/mol
Density:1,72 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together