3-Methyl-1-(4-nitrophenyl)-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-ylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:852690-94-9
:
Formula:C₁₇H₁₃F₃N₄O₂
MDL Number:MFCD03791150
:362.31 g/mol
Melting Pt:163…165 °C
Storage Temperature:Ambient
Frequently Bought Together