3-Fluoro-DL-phenylglycine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7292-74-2
:
Formula:C₈H₈FNO₂
MDL Number:MFCD00049325
:169.16 g/mol
Melting Pt:300 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together