2,4,6-Trichloroaniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:634-93-5
:
Formula:C₆H₄Cl₃N
MDL Number:MFCD00007663
:196.46 g/mol
Boiling Pt:262 °C
Melting Pt:73…75 °C
EINECS:211-219-8
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together